Skip to Content
Merck
All Photos(1)

Documents

W4270

Sigma-Aldrich

WKYMVM trifluoroacetate salt

≥98% (HPLC)

Synonym(s):

Trp-Lys-Tyr-Met-Val-L-Met NH2 trifluoroacetate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H61N9O7S2 · xC2HF3O2
CAS Number:
Molecular Weight:
856.11 (free base basis)
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.32

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

H2O: >2 mg/mL

shipped in

wet ice

storage temp.

−20°C

SMILES string

OC(=O)C(F)(F)F.CSCC[C@H](NC(=O)[C@@H](NC(=O)[C@H](CCSC)NC(=O)[C@H](Cc1ccc(O)cc1)NC(=O)[C@H](CCCCN)NC(=O)[C@@H](N)Cc2c[nH]c3ccccc23)C(C)C)C(N)=O

InChI

1S/C41H61N9O7S2.C2HF3O2/c1-24(2)35(41(57)46-31(36(44)52)16-19-58-3)50-39(55)33(17-20-59-4)48-40(56)34(21-25-12-14-27(51)15-13-25)49-38(54)32(11-7-8-18-42)47-37(53)29(43)22-26-23-45-30-10-6-5-9-28(26)30;3-2(4,5)1(6)7/h5-6,9-10,12-15,23-24,29,31-35,45,51H,7-8,11,16-22,42-43H2,1-4H3,(H2,44,52)(H,46,57)(H,47,53)(H,48,56)(H,49,54)(H,50,55);(H,6,7)/t29-,31-,32-,33-,34-,35-;/m0./s1

InChI key

MYVYQWVNXDVTBW-CZPFDPCBSA-N

Amino Acid Sequence

Trp-Lys-Tyr-Met-Val-L-Met NH2

Application

WKYMVM trifluoroacetate salt was used as a test compound for investigating the mechanism of regulation of lipoprotein-associated phospholipase A2 (Lp-PLA2).

Biochem/physiol Actions

WKYMVM is a peptide agonist of formyl peptide receptors. WKYMVM binds to and stimulates FPRL family members 1, 2 with EC50 values of 2 and 80 nM, respectively. The peptide is a potent activator of neutrophils.
WKYMVM is an agonist and potent activator of Formyl peptide receptor like-1 (FPRL1). WKYMVM stimulates FPRL1 and hence attenuates the function of the chemokine receptors CCR5 and CXCR4. WKYMVM activates human eosinophils to induce superoxide production through Phosphoinositide 3-Kinase-Mediated ERK1/2 pathway.

Preparation Note

WKYMVM trifluoroacetate salt dissolves in water at a concentration >2 mg/mL to yield a clear solution.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Myeong Heon Shin et al.
International archives of allergy and immunology, 137 Suppl 1, 21-26 (2005-06-11)
Eosinophils play a key role in allergic inflammation and parasitic infections. The synthetic peptide, Trp-Lys-Tyr-Met-Val-D-Met (WKYMVm), has been previously shown to activate eosinophils and thus to enhance respiratory burst through the formyl peptide receptors. This study was undertaken to determine
B Q Li et al.
Blood, 97(10), 2941-2947 (2001-05-09)
The G protein-coupled 7 transmembrane (STM) chemoattractant receptors can be inactivated by heterologous desensitization. Earlier work showed that formly peptide receptor-like 1 (FPRL1), an STM receptor with low affinity for the bacterial chemotactic peptide formyl-methionyl-leucyl-phenylalamine (fMLF), is activated by peptide
Bo Li et al.
Atherosclerosis, 228(2), 370-379 (2013-04-30)
Although lipoprotein-associated phospholipase A2 (Lp-PLA2) has been regarded as a biomarker and a causative agent for acute coronary events recently, the mechanism of the regulation of Lp-PLA2 has not been fully elucidated yet. This study was aimed to investigate the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service