Skip to Content
Merck
All Photos(1)

Key Documents

U5633

Sigma-Aldrich

Ubiquitin Activating Enzyme (E1) human

recombinant, expressed in baculovirus infected Sf21 cells

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
R$4,577.00

R$4,577.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μG
R$4,577.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.56

R$4,577.00


Check Cart for Availability

Request a Bulk Order

biological source

human

Quality Level

recombinant

expressed in baculovirus infected Sf21 cells

Assay

≥95% (SDS-GE)

form

solution

UniProt accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... UBA1(7317)

General description

Ubiquitin Activating Enzyme (E1) is responsible for the first step in ubiquitin-protein isopeptide bond formation. E1 catalyzes the activation of the C-terminal carboxyl group of ubiquitin by forming a high-energy thioester bond in an ATP-dependent manner.

Physical form

Solution in 50 mM HEPES, pH 8.0, 50 mM NaCl and 1mM TCEP

Other Notes

Manufactured for Sigma by Boston Biochem. Inc.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chunmei Yang et al.
International journal of biological sciences, 16(6), 1059-1070 (2020-03-07)
Cullin 4A and 4B (CUL4A and 4B) function as oncogenes in colorectal cancer (CRC) cells. Both of them conservatively associate with DNA damage-binding protein 1 (DDB1) and DDB1-CUL4-associated factor 4 (DCAF4) to form Cullin-RING E3 ligases known as CRL4DCAF4, which

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service