Skip to Content
Merck
All Photos(1)

Key Documents

U3885

Sigma-Aldrich

UK-78282 monohydrochloride

≥98% (HPLC)

Synonym(s):

4-[(Diphenylmethoxy)methyl]-1-[3-(4-methoxyphenyl)propyl]-piperidine monohydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$1,421.00

R$1,421.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
R$1,421.00

About This Item

Empirical Formula (Hill Notation):
C29H35NO2HCl
CAS Number:
Molecular Weight:
466.05
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

R$1,421.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

solid

storage condition

desiccated

color

white to off-white

solubility

DMSO: >10 mg/mL

shipped in

wet ice

storage temp.

−20°C

SMILES string

Cl.COc1ccc(CCCN2CCC(CC2)COC(c3ccccc3)c4ccccc4)cc1

InChI

1S/C29H35NO2.ClH/c1-31-28-16-14-24(15-17-28)9-8-20-30-21-18-25(19-22-30)23-32-29(26-10-4-2-5-11-26)27-12-6-3-7-13-27;/h2-7,10-17,25,29H,8-9,18-23H2,1H3;1H

InChI key

ZZSQARULZYQMIG-UHFFFAOYSA-N

Biochem/physiol Actions

UK-78282 blocks both Kv1.3 and Kv1.4. Kv1.3 and Kv1.4 are members of the Shaker family of voltage-gated potassium channels.
UK-78282 blocks both Kv1.3 and Kv1.4. Kv1.3 and Kv1.4 are members of the Shaker family of voltage-gated potassium channels. Kv1.3 is expressed in brain and in effector memory T (Tem) cells, and Kv1.4 is expressed in brain. Both channels are involved in setting the membrane potential of neurons. In addition, Kv1.3 maintains the membrane potential for T memory cells and is up-regulated upon T cell activation, contributing to multiple immune processes and disorders. Blockers of Kv1.3 have long been sought for therapeutic benefit, and they are also valuable tools for studying the immune system. Selective blockers of Kv1.4 are desired, as there are few if any currently available. UK-78282 is valuable for immune system studies, where the absence of Kv1.4 makes it functionally selective for Kv1.3, and it has shown suppression of T cell activation. UK-78282 is also valuable for the study of Kv1.4 function.

Features and Benefits

This compound is featured on the Potassium Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Preparation Note

UK-78282 monohydrochloride dissolves in DMSO at a concentration >10 mg/ml.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ruxandra Anton et al.
International journal of molecular sciences, 22(4) (2021-03-07)
(1) Background: As membrane channels contribute to different cell functions, understanding the underlying mechanisms becomes extremely important. A large number of neuronal channels have been investigated, however, less studied are the channels expressed in the glia population, particularly in microglia.

Articles

We offer many products related to potassium channels for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service