Skip to Content
Merck
All Photos(1)

Key Documents

U2258

Sigma-Aldrich

Ubiquitin-7-amido-4-methylcoumarin

≥95% (HPLC), solution (supplied in DMSO)

Synonym(s):

Ubiquitin-AMC

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
R$7,360.00
5 X 50 μG
R$17,916.00

R$7,360.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 μG
R$7,360.00
5 X 50 μG
R$17,916.00

About This Item

MDL number:
UNSPSC Code:
12352204
NACRES:
NA.32

R$7,360.00


Check Cart for Availability

Request a Bulk Order

Product Name

Ubiquitin-7-amido-4-methylcoumarin, solution (supplied in DMSO)

Assay

≥95% (HPLC)

Quality Level

form

solution (supplied in DMSO)

shipped in

dry ice

storage temp.

−70°C

General description

Manufactured for Sigma by Boston Biochem. Inc.
Ubiquitin-7-amido-4-methylcoumarin (Ub-AMC) is a fluorogenic Ub-base probe. It consists of AMC conjugated to the glycine residue of ubiquitin.[1] Ub-AMC is a fluorogenic substrate for ubiquitin C-terminal hydrolase-L3 (UCH-L3) as well as other UCH isozymes.[2]

Application

Ubiquitin-7-amido-4-methylcoumarin (Ub-AMC) has been used as a fluorogenic substrate in deubiquitinase assay buffer.[3]

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F

Flash Point(C)

87 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Efficient chemical synthesis for the analogue of ubiquitin-based probe Ub--AMC with native bioactivity
Xu L, et al.
Royal Society of Chemistry Advances, 6(53), 47926-47930 (2016)
Kazunori Hirayama et al.
Bioorganic & medicinal chemistry, 15(21), 6810-6818 (2007-09-01)
UCH-L3 (ubiquitin C-terminal hydrolase-L3) is a de-ubiquitinating enzyme that is a component of the ubiquitin-proteasome system and known to be involved in programmed cell death. A previous study of high-throughput drug screening identified an isatin derivative as a UCH-L3 inhibitor.
Sehyoun Yoon et al.
Neuron, 105(3), 506-521 (2019-12-10)
Variants in the ANK3 gene encoding ankyrin-G are associated with neurodevelopmental disorders, including intellectual disability, autism, schizophrenia, and bipolar disorder. However, no upstream regulators of ankyrin-G at synapses are known. Here, we show that ankyrin-G interacts with Usp9X, a neurodevelopmental-disorder-associated deubiquitinase (DUB).

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service