Skip to Content
Merck
All Photos(1)

Key Documents

U111

Sigma-Aldrich

(−)-trans-(1S,2S)-U-50488 hydrochloride hydrate

solid, ≥98% (HPLC)

Synonym(s):

trans-(1S,2S)-3,4-Dichloro-N-methyl-N-[2-(1-pyrrolidinyl)cyclohexyl]benzeneacetamide hydrochloride hydrate

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$1,134.00
25 MG
R$3,393.00
100 MG
R$11,335.00

R$1,134.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
R$1,134.00
25 MG
R$3,393.00
100 MG
R$11,335.00

About This Item

Empirical Formula (Hill Notation):
C19H26Cl2N2O · HCl · xH2O
CAS Number:
Molecular Weight:
405.79 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

R$1,134.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

solid

optical activity

[α]22/D -39.0 to -32.0°, c = 0.5 in methanol(lit.)

storage condition

desiccated

color

white

solubility

H2O: 13 mg/mL

storage temp.

2-8°C

SMILES string

O.Cl.CN([C@H]1CCCC[C@@H]1N2CCCC2)C(=O)Cc3ccc(Cl)c(Cl)c3

InChI

1S/C19H26Cl2N2O.ClH.H2O/c1-22(19(24)13-14-8-9-15(20)16(21)12-14)17-6-2-3-7-18(17)23-10-4-5-11-23;;/h8-9,12,17-18H,2-7,10-11,13H2,1H3;1H;1H2/t17-,18-;;/m0../s1

InChI key

UWPRPWMHQDPOFR-MPGISEFESA-N

Gene Information

human ... OPRK1(4986)

Application

(−)-trans-(1S,2S)-U-50488 hydrochloride hydrate (U-50488) has been used as a κ-opioid receptor (KOR) agonist has been used to study its effect on the increase of tidal volumes in adult red-eared slider turtles (Trachemys scripta)[1].

Biochem/physiol Actions

Potent κ opioid receptor agonist; more potent enantiomer of (±)-trans-U-50488.

Preparation Note

(−)-trans-(1S,2S)-U-50488 hydrochloride hydrate is soluble in water at 13 mg/ml.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stephen M Johnson et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 295(5), R1599-R1612 (2008-09-12)
For ectothermic vertebrates, such as reptiles, the effects of opioid receptor subtype activation on breathing are poorly understood. On the basis of previous studies on mammals and lampreys, we hypothesized that mu- and delta-opioid receptor (MOR and DOR, respectively) activation
Stereospecific inhibition of gastro-intestinal transit by κ-opioid agonists in mice.
Ramabadran, et al.
European Journal of Pharmacology, 155, 329-329 (1991)
D D Taub et al.
Proceedings of the National Academy of Sciences of the United States of America, 88(2), 360-364 (1991-01-15)
Opioids and opioid peptides have been shown by numerous laboratories to modulate various parameters of the immune response, but little attention has been given to the type of opioid receptor that might be involved. This study focuses on the in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service