TMO95040452
Nunc® 96 DeepWell™ plate, non-treated
untreated, polystyrene, non-sterile, without lid, natural, case of 50 ea
Synonym(s):
96 deepwell culture plate, 96 deepwell plate, sample processing plate, sample storage plate
Select a Size
R$4,074.00
Select a Size
About This Item
polypropylene
polystyrene
R$4,074.00
Recommended Products
material
natural
polypropylene
polystyrene
sterility
non-sterile
feature
lid: no
packaging
case of 50 ea
manufacturer/tradename
Nunc 95040452
plate L × W
128 mm × 86 mm
size
96 well
color
natural
suitability
suitable for (sample collection, storage, combinatorial chemistry and library applications)
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- Unique low design with shared well technology for increased well volume
- 1.3 and 2.0 mL volume capacity
- Optimum storage capacity and improved mixing
- Round well shape reduces liquid retention
- Well cap available for safe storage – minimal protrusion into each well
- Ideal for sample collection, storage, combinatorial chemistry and library applications
- Resistant to most chemicals, solvents and alcohols used in combinatorial chemistry
- Widely used for bacterial and yeast growth
- Easy-to-use with automatic sample handling instruments
- Alpha-numeric grid for quick sample identification
- Ideal for sample collection, storage, and combinatorial chemistry library applications
- Sealable with well caps and sealing tape
- Ideal as collection plates for Nunc Filter Plates
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service