Skip to Content
Merck
All Photos(1)

Key Documents

TMO362800-0020

Nalgene® Natural Micro Packaging Vial

polypropylene copolymer, vial volume 2.0 mL, conical bottom, self-standing, external thread, untreated, case of 1000 ea

Synonym(s):

Nalgene vial, plastic vial

Sign Into View Organizational & Contract Pricing

Select a Size

1000 EACH
R$1,741.00

R$1,741.00


Check Cart for Availability


Select a Size

Change View
1000 EACH
R$1,741.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.11

R$1,741.00


Check Cart for Availability

material

conical bottom
natural
polypropylene copolymer
self-standing

packaging

case of 1000 ea

manufacturer/tradename

Nalgene 362800-0020

technique(s)

PCR: suitable

vial volume

2.0 mL

Looking for similar products? Visit Product Comparison Guide

General description

Completely recover vial contents with skirted, conical-interior Thermo Scientific Nalgene Natural PPCO Micro Packaging Vials, molded from high-purity, noncytotoxic, nonpyrogenic, low-metal content polypropylene copolymer. Vials and closures (listed separately) are pressure-tested together at 7.5 psig for safe air shipment and can be centrifuged at 13,000 x g.

Features and Benefits

  • USP Class VI compliant
  • Ensure sample integrity with natural, high-purity polypropylene copolymer that is noncytotoxic, nonpyrogenic, and low in metal content, supporting reliable experimental outcomes
  • Colored closures packaged separately
  • Nonsterile
  • Leakproof† when used with Nalgene closures
  • Facilitate complete recovery of vial contents with a conical interior and skirted design, minimizing sample waste and ensuring accurate experimental replication.
  • Safe air shipment with closures pressure-tested at 7.5 psig, mitigating risks during transport and preserving sample quality.
  • Enhanced versatility and durability, enabling centrifugation at speeds up to 13,000 x g, meeting demanding experimental requirements.

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service