Documents
TMO312006-9125
Nalgene® diagnostic bottle
natural polypropylene copolymer, volume 4 mL, case of 2000 ea
Synonym(s):
PPCO bottles, small volume bottle with closure
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
material
natural polypropylene copolymer
packaging
case of 2000 ea
manufacturer/tradename
Nalgene 312006-9125
capacity
4 mL
volume
4 mL
General description
Sample, store and ship small volume, valuable reagents. Thermo Scientific Nalgene Natural PPCO Diagnostic Bottles with Closure are durable, autoclavable and used in test kits and many small-volume applications. These natural, translucent polypropylene copolymer bottles feature uniform walls resistant to splitting or puncturing. Bottles include linerless polypropylene closures that ensure leakproof† performance without a liner that can wrinkle, cause leaks and contaminate your solutions.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Visit the Document Library
Related Content
FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.