Key Documents
TMO2401-0125
Nalgene® economy wash bottle
low-density polyethylene, volume 125 mL, unlabelled, case of 48 ea
Synonym(s):
LDPE flush bottle, LDPE wash bottle
Select a Size
48 EACH
R$2,267.00
R$2,267.00
Check Cart for Availability
Select a Size
Change View
48 EACH
R$2,267.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
R$2,267.00
Check Cart for Availability
material
low-density polyethylene
packaging
case of 48 ea
manufacturer/tradename
Nalgene 2401-0125
capacity
125 mL
volume
125 mL
General description
Prevent separation when squeezing Thermo Scientific Nalgene LDPE Economy Wash Bottles, featuring a tight-fitting draw tube and closure. These low density polyethylene bottles are designed to dispense through the top with a closure and stem molded in one piece for uniform leakproof† service. Cut back tip to increase flow.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)