Key Documents
TMO163371
Nunc® cell culture treated flask with solid cap
polystyrene, flask capacity 7 mL, Nunclon® Delta surface, sterile, case of 160 ea
Synonym(s):
cell culture flask, treated flask
Select a Size
160 EACH
R$3,662.00
R$3,662.00
Check Cart for Availability
Select a Size
Change View
160 EACH
R$3,662.00
About This Item
UNSPSC Code:
41161502
NACRES:
NB.11
R$3,662.00
Check Cart for Availability
material
polystyrene
sterility
sterile
packaging
case of 160 ea
manufacturer/tradename
Nunc 163371
flask capacity
7 mL
General description
Achieve maximum adhesion for a broad range of cell types with Thermo Scientific Nunc Cell Culture Treated Flasks with Filter Caps. These flasks receive the proprietary Nunclon Delta surface treatment to ensure consistent cell growth. The filter caps allow constant airflow and minimize chances of contamination.
Legal Information
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Nunclon is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)