Key Documents
TMO140656
Nunc® Cell Culture Inserts in 12-well Nunclon Delta surface plate
cell culture treated, polycarbonate, sterile, pore size 8.0 μm, case of 48 ea (supplied in 4 x 12 well plates)
Synonym(s):
multidish membrane insert, treated membrane insert
Select a Size
48 EACH
R$3,982.00
R$3,982.00
Estimated to ship onJune 01, 2025
Select a Size
Change View
48 EACH
R$3,982.00
About This Item
UNSPSC Code:
41122102
NACRES:
NB.13
R$3,982.00
Estimated to ship onJune 01, 2025
material
polycarbonate
sterility
sterile
packaging
case of 48 ea (supplied in 4 x 12 well plates)
manufacturer/tradename
Nunc 140656
pore size
8.0 μm pore size
binding type
cell culture treated
General description
Cultivate most cell types without matrix coating when using Thermo Scientific Nunc Polycarbonate Membrane Inserts in Multidishes.
Legal Information
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)