Skip to Content
Merck
All Photos(1)

Key Documents

T9823

Sigma-Aldrich

Tetracycline hydrochloride

Biotechnology Performance Certified, suitable for cell culture, ≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H24N2O8 · HCl
CAS Number:
Molecular Weight:
480.90
Beilstein:
3844873
EC Number:
MDL number:
UNSPSC Code:
51101500
PubChem Substance ID:

grade

Biotechnology Performance Certified

Assay

≥95%

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

mp

220-223 °C (lit.)

solubility

H2O: soluble 50 mg/mL

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

Mode of action

protein synthesis | interferes

storage temp.

−20°C

SMILES string

Cl.CN(C)[C@H]1[C@@H]2C[C@H]3C(=C(O)[C@]2(O)C(=O)C(C(N)=O)=C1O)C(=O)c4c(O)cccc4[C@@]3(C)O

InChI

1S/C22H24N2O8.ClH/c1-21(31)8-5-4-6-11(25)12(8)16(26)13-9(21)7-10-15(24(2)3)17(27)14(20(23)30)19(29)22(10,32)18(13)28;/h4-6,9-10,15,25,27-28,31-32H,7H2,1-3H3,(H2,23,30);1H/t9-,10-,15-,21+,22-;/m0./s1

InChI key

XMEVHPAGJVLHIG-FMZCEJRJSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: tetracycline

Application

Tetracycline is a broad spectrum polyketide antibiotic. It is used clinically to treat bacterial infections such as Rocky Mountain spotted fever, typhus fever, tick fevers, Q fever and Brill-Zinsser disease. It is used to treat upper respiratory infections and acne[1]. It is used to study multidrug resistance[2] as well as potential side effects such as acute pancreatitis[3]. It has been used in target and resistance based mechanistic studies of novel antibiotics[4].

Biochem/physiol Actions

Mode of Action: Tetracycline passively diffuses through proin channels in the cell membrane, binding to 30S ribosomes and inhibits protein synthesis by preventing access of aminoacyl tRNA to the acceptor site on the mRNA-ribosome complex. It also binds to the bacterial 50S ribosomal subunit, altering the membrane and causing intracellular components to leak from bacterial cells. The inhibitory effects can be reversed by washing, suggesting that it is the reversibly bound antibiotic, and not the irreversibly bound drug, that is responsible for antibacterial action.

Mode of Resistance: The effects are inactivated via a loss of cell wall permeability.

Antimicrobial spectrum: Includes a wide range of antimicrobial activity against gram-positive and gram-negative bacteria.
Tetracycline inhibits bacterial protein synthesis elongation at the level of aminoacyl-tRNA binding to the 30S ribosome. Tetracycline passively diffuses through porin channels in the bacterial cell membrane. It also binds to the bacterial 50S ribosomal subunit which may alter the cytoplasmic membrane, causing intracellular components to leak from bacterial cells[1]. Mode of resistance is loss of cell wall permeability.

Signal Word

Warning

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Trudy H Grossman et al.
Antimicrobial agents and chemotherapy, 56(5), 2559-2564 (2012-02-23)
TP-434 is a novel, broad-spectrum fluorocycline antibiotic with activity against bacteria expressing major antibiotic resistance mechanisms, including tetracycline-specific efflux and ribosomal protection. The mechanism of action of TP-434 was assessed using both cell-based and in vitro assays. In Escherichia coli
Ricardo E de Cristóbal et al.
The Journal of antimicrobial chemotherapy, 58(1), 31-36 (2006-05-12)
Starting from the observation that Escherichia coli tolC mutations severely reduced the high-level resistance to tetracycline afforded by Tn10- and plasmid-encoded Tet(A) pumps, we studied the mechanism of this susceptibility. The MIC of tetracycline for MC4100 tolC::Tn10 and several tolC
Increased risk of acute pancreatitis among tetracycline users in a Swedish population-based case-control study
Rickard Ljung, Jesper Lagergren, et al.
Gut, 10.1136, 300949-300949 (2011)
Biyang Deng et al.
Food chemistry, 134(4), 2350-2354 (2013-02-28)
A novel and sensitive determination for tetracycline (TC) in crucian carp muscle was developed by using capillary electrophoresis coupled with electrochemiluminescence (ECL) detection. The conditions affected separation and detection were examined in detail. The linearity range of TC concentration was
Leslie Cuthbertson et al.
Chemistry & biology, 20(2), 232-240 (2013-02-27)
The ligands that interact with the vast majority of small-molecule binding transcription factors are unknown, a significant gap in our understanding of sensory perception by cells. TetR-family regulators (TFRs) are found in most prokaryotes and are involved in regulating virtually

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service