T8141
Titanium(IV) oxide, anatase
≥99%
Synonym(s):
Titanium dioxide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
TiO2
CAS Number:
Molecular Weight:
79.87
EC Number:
MDL number:
UNSPSC Code:
12352300
Recommended Products
Assay
≥99%
form
powder
particle size
−325 mesh
mp
1825 °C (lit.)
density
3.9 g/mL at 25 °C (lit.)
SMILES string
O=[Ti]=O
InChI
1S/2O.Ti
InChI key
GWEVSGVZZGPLCZ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
13 - Non Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Ana R Ribeiro et al.
Nanoscale, 9(30), 10684-10693 (2017-06-28)
While titanium is the metal of choice for most prosthetics and inner body devices due to its superior biocompatibility, the discovery of Ti-containing species in the adjacent tissue as a result of wear and corrosion has been associated with autoimmune
Saud Alarifi et al.
International journal of nanomedicine, 8, 3937-3943 (2013-10-22)
Titanium dioxide (TiO₂) nanoparticles are among the top five nanoparticles used in consumer products, paints, and pharmaceutical preparations. Given that exposure to such nanoparticles is mainly via the skin and inhalation, the present study was conducted in male Wistar albino
Zahra Zahra et al.
Journal of agricultural and food chemistry, 66(32), 8522-8529 (2018-07-18)
This study aimed to improve the production of phycobiliproteins using TiO2 nanoparticles (NPs) in Synechocystis sp. PCC 6803. The growth characteristics of Synechocystis cells were not affected by TiO2 NPs treatment, but this treatment increased the chlorophyll content significantly by
Ewa Borowska et al.
Water, air, and soil pollution, 226(5), 151-151 (2015-05-12)
Iodinated contrast media (ICM), which are used for radiological visualization of human tissue and cardiovascular system, are poorly biodegradable; hence, new methods of their removal are sought. In this study, the effectiveness of selected X-ray ICM removal by means of
Swati Srivastava et al.
Journal of environmental sciences (China), 55, 11-19 (2017-05-10)
The objective of this study was to understand toxicity of mixture of nanoparticles (NPs) (ZnO and TiO
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service