Skip to Content
Merck
All Photos(1)

Key Documents

T5537

Sigma-Aldrich

Triethylene glycol dimethacrylate

~95% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=C(CH3)COO(CH2CH2O)3COC(CH3)=CH2
CAS Number:
Molecular Weight:
286.32
Beilstein:
1797351
EC Number:
MDL number:
UNSPSC Code:
12171500

Assay

~95% (GC)

contains

~80 ppm hydroquinone as inhibitor

refractive index

n20/D 1.461 (lit.)

bp

170-172 °C/5 mmHg (lit.)

density

1.092 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(=C)C(=O)OCCOCCOCCOC(=O)C(C)=C

InChI

1S/C14H22O6/c1-11(2)13(15)19-9-7-17-5-6-18-8-10-20-14(16)12(3)4/h1,3,5-10H2,2,4H3

InChI key

HWSSEYVMGDIFMH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

332.6 °F - closed cup

Flash Point(C)

167 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service