Skip to Content
Merck
All Photos(1)

Key Documents

T3889

Sigma-Aldrich

Tetracycline phosphate complex

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
UNSPSC Code:
51284046

form

powder

color

yellow to yellow with an orange cast

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

Mode of action

protein synthesis | interferes

storage temp.

−20°C

General description

Chemical structure: tetracycline

Application

Tetracycline is a broad spectrum polyketide antibiotic. It is used clinically to treat bacterial infections such as Rocky Mountain spotted fever, typhus fever, tick fevers, Q fever and Brill-Zinsser disease. It is used to treat upper respiratory infections and acne. It is used to study multidrug resistance[1] as well as potential side effects such as acute pancreatitis[2].

Biochem/physiol Actions

Tetracycline inhibits bacterial protein synthesis elongation at the level of aminoacyl-tRNA binding to the 30S ribosome. Tetracycline passively diffuses through porin channels in the bacterial cell membrane. It also binds to the bacterial 50S ribosomal subunit which may alter the cytoplasmic membrane, causing intracellular components to leak from bacterial cells[3].

Packaging

25G

Storage and Stability

Keep container tightly closed in a dry and well-ventilated place. Storage class (TRGS 510): Non-combustible, acute toxic hazardous, acute toxic Cat. 3 / toxic hazardous materials or hazardous materials causing chronic effects.

Other Notes

Keep container tightly closed in a dry and well-ventilated place. Storage class (TRGS 510): Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Increased risk of acute pancreatitis among tetracycline users in a Swedish population-based case-control study
Rickard Ljung, Jesper Lagergren, et al.
Gut, 10.1136, 300949-300949 (2011)
Ricardo E de Cristóbal et al.
The Journal of antimicrobial chemotherapy, 58(1), 31-36 (2006-05-12)
Starting from the observation that Escherichia coli tolC mutations severely reduced the high-level resistance to tetracycline afforded by Tn10- and plasmid-encoded Tet(A) pumps, we studied the mechanism of this susceptibility. The MIC of tetracycline for MC4100 tolC::Tn10 and several tolC
Craig Knox et al.
Nucleic acids research, 39(Database issue), D1035-D1041 (2010-11-10)
DrugBank (http://www.drugbank.ca) is a richly annotated database of drug and drug target information. It contains extensive data on the nomenclature, ontology, chemistry, structure, function, action, pharmacology, pharmacokinetics, metabolism and pharmaceutical properties of both small molecule and large molecule (biotech) drugs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service