Skip to Content
Merck
All Photos(1)

Key Documents

T3512

Sigma-Aldrich

Thymidine 3′-monophosphate sodium salt

99% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N2O8PNa
CAS Number:
Molecular Weight:
344.19
UNSPSC Code:
41106305
PubChem Substance ID:

Assay

99% (HPLC)

storage temp.

−20°C

SMILES string

O=C1N([C@@H]2O[C@H](CO)[C@@H](CP(O)(O)=O)C2)C=C(C)C(N1)=O.[CH4]

InChI

1S/C10H15N2O8P.Na.H/c1-5-3-12(10(15)11-9(5)14)8-2-6(7(4-13)19-8)20-21(16,17)18;;/h3,6-8,13H,2,4H2,1H3,(H,11,14,15)(H2,16,17,18);;

InChI key

KTAXSUHEUYNIOC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Preparation Note

Enzymatically prepared.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service