T2826
TargeTron™ Vector pACD4K-C-loxP
Expression Vector for Bacterial Gene Knockout
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352200
Recommended Products
shipped in
wet ice
storage temp.
−20°C
General description
The TargeTron Vector pACD4K-C-loxP is a 7,745bp E. coli expression vector for use with the TargeTron Gene Knockout System (TA0100). The vector is linearized at the HindIII and BsrGI sites. This vector differs from the original pACD4K-C vector (included in the TargeTron Gene Knockout System) in that it has loxP sites flanking each end of the kanamycin ORF that allow for removal of the kanamycin resistance marker via Cre-loxP mediated recombination. After removal of the kanamycin resistance marker, sequential knockouts can be generated.
Other Notes
For more information and to view applications data, please visit www.sigma-aldrich.com/targetron.
Legal Information
TargeTron is a trademark of InGex, LLC
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service