Skip to Content
Merck
All Photos(1)

Key Documents

T2293

Nalgene® centrifuge tubes, Oak Ridge Style 3114

capacity 30 mL, pack of 2 ea

Synonym(s):

High-Speed ​​Centrifuge Tubes, Nalgene tubes

Sign Into View Organizational & Contract Pricing

Select a Size

2 EA
R$1,915.00

R$1,915.00


Check Cart for Availability


Select a Size

Change View
2 EA
R$1,915.00

About This Item

UNSPSC Code:
41121703
eCl@ss:
32119210
NACRES:
NB.11

R$1,915.00


Check Cart for Availability

material

FEP
Tefzel® closure
round bottom
screw top cap

sterility

non-sterile

feature

RCF 50,000 × g with sealing cap assembly

packaging

pack of 2 ea

manufacturer/tradename

Nalgene® 3114-0030

capacity

30 mL

Looking for similar products? Visit Product Comparison Guide

General description

[TM=“Nalgene”] Oak Ridge high-speed centrifuge tubes made with [TM=“PTFE”] fluoropolymer and closure made with [TM=“Tefzel”] can be used to perform high-speed centrifugation of aggressive chemicals. These tubes and closures are specially designed for chloroform and phenol extractions.

Features and Benefits

  • Capable of withstanding temperatures ranging from -100°C to +150°C (-148°F to +302°F).
  • Leakproof closures prevent material loss under normal use for most spins <10,000 × g.
  • When used with Nalgene sealing cap assemblies, the 30mL tubes can handle centrifugal forces up to 50,000 x g.
  • Autoclavable, biohazard-safe, and leakproof when utilized with the provided Nalgene closure or PTFE FEP.

Other Notes

Tubes must be filled to at least 100% of their total capacity to ensure proper performance.

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Tefzel is a registered trademark of E. I. du Pont de Nemours and Company

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service