T2068
Greiner centrifuge tubes
capacity 50 mL, diam. × L 30 mm × 115 mm, conical bottom (with skirt)
Synonym(s):
centrifuge tubes, culture tubes, plastic centrifuge tube, plastic culture tube, Greiner centrifuge tubes
Select a Size
R$3,452.00
Select a Size
About This Item
R$3,452.00
Recommended Products
material
cap
conical bottom (with skirt)
polypropylene tube
sterility
sterile
feature
RCF 3,200 × g
graduations
packaging
pack of 25
case of 450
manufacturer/tradename
Greiner 210261
capacity
50 mL
diam. × L
30 mm × 115 mm
color
blue cap
colorless tube
Looking for similar products? Visit Product Comparison Guide
General description
High quality, crystal-clear polypropylene centrifuge tubes. Centrifuge tubes can be safely centrifuged to approximately 3,200 × g (50 mL size) and 3,500 × g (15 mL size) depending on the specific gravity of the solution being centrifuged and the type of rotor.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
A complete workflow for estimating the total polyphenol content of apple and banana samples, with gallic acid standard addition curve protocol.
Discover the detection of milk adulteration by whey using spectrophotometric estimation of sialic acid in three different milk brands. Learn more about this essential technique in our application note.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service