Key Documents
T1906
Sigma-Aldrich
Ultramicro pipette tips
Microcapillary, round tip, maximum volume 200 μL, non-sterile
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
material
colorless
polypropylene
sterility
non-sterile
feature
barrier: no
packaging
pack of 200 ea (Trays)
manufacturer/tradename
Sigma-Aldrich
maximum volume
200 μL
General description
For loading tube gels and slab gels 0.6 mm or thicker. Mark at 5 μl gives visual confirmation of proper pipet operation. Each tray contains 10 × 20 tips
Low capacity improves accuracy with small sample volumes by reducing dead space.
Other Notes
Browse all pipette tips
Choose from one of the most recent versions:
Certificates of Analysis (COA)