Skip to Content
Merck
All Photos(1)

Key Documents

T1081

Sigma-Aldrich

TLQP-21 human

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
R$2,443.00
1 MG
R$4,654.00

R$2,443.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Select a Size

Change View
500 μG
R$2,443.00
1 MG
R$4,654.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

R$2,443.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

form

lyophilized powder

Quality Level

color

white

solubility

deionized water: soluble

shipped in

dry ice

storage temp.

−20°C

Amino Acid Sequence

H-THR-LEU-GLN-PRO-PRO-SER-ALA-LEU-ARG-ARG-ARG-HIS-TYR-HIS-HIS-ALA-LEU-PRO-PRO-SER-ARG-OH

General description

TLQP-21 peptide is a derivative of VGF and is expressed at higher levels in the brain.[1]

Biochem/physiol Actions

TLQP-21 peptide is involved in the central and peripheral nervous system activities such as regulating reproductive and circadian behaviors, feeding, and body weight. It is also involved in regulating neuropathic pain and peripheral adiposity.[1] TLQP-21 plays a role in stimulating the secretion of insulin and prevents apoptosis of pancreatic cells.[2] It also activates the complement C3a receptor-1 (C3aR1) in the brain.[1]

Physical form

Lyophilized from 0.1% aqueous TFA

Other Notes

Lyophilized from 0.1% TFA in H2O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei Zhang et al.
PloS one, 8(11), e79760-e79760 (2013-11-28)
Hyperglycemia causes oxidative stress that could damage vascular endothelial cells, leading to cardiovascular complications. The Vgf gene was identified as a nerve growth factor-responsive gene, and its protein product, VGF, is characterized by the presence of partially cleaved products. One
C Severini et al.
British journal of pharmacology, 157(6), 984-993 (2009-05-27)
Vgf gene expression has been detected in various endocrine and neuronal cells in the gastrointestinal tract. In this study we investigated the pharmacological activity of different VGF-derived peptides. Among these, TLQP-21, corresponding to the 556-576 fragment of the protein was
Farida El Gaamouch et al.
Molecular neurodegeneration, 15(1), 4-4 (2020-01-12)
Multiomic studies by several groups in the NIH Accelerating Medicines Partnership for Alzheimer's Disease (AMP-AD) identified VGF as a major driver of Alzheimer's disease (AD), also finding that reduced VGF levels correlate with mean amyloid plaque density, Clinical Dementia Rating
A Bartolomucci et al.
Proceedings of the National Academy of Sciences of the United States of America, 103(39), 14584-14589 (2006-09-20)
The vgf gene has been identified as an energy homeostasis regulator. Vgf encodes a 617-aa precursor protein that is processed to yield an incompletely characterized panel of neuropeptides. Until now, it was an unproved assumption that VGF-derived peptides could regulate
Huan Liu et al.
Autophagy, 16(12), 2140-2155 (2020-01-22)
Mesenchymal stem cell (MSC) transplantation has been widely applied as a potential therapeutic for multiple diseases. However, the underlying therapeutic mechanisms are not fully understood, especially the paradox between the low survival rate of transplanted cells and the beneficial therapeutic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service