Skip to Content
Merck
All Photos(1)

Documents

T0893

Sigma-Aldrich

cis-9,trans-12-Tetradecadienyl acetate

≥85% (GC)

Synonym(s):

(Z,E)-9,12-TDDA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H28O2
CAS Number:
Molecular Weight:
252.39
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

Assay

≥85% (GC)

form

liquid

functional group

ester

shipped in

ambient

storage temp.

−20°C

SMILES string

C\C=C\C\C=C/CCCCCCCCOC(C)=O

InChI

1S/C16H28O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-18-16(2)17/h3-4,6-7H,5,8-15H2,1-2H3/b4-3+,7-6-

InChI key

ZZGJZGSVLNSDPG-FDTUMDBZSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Pheromone for Anagasta kuhniella, Cadra elutella, Cadra figulilella, Cadra cautella, Plodia interpunctella, Spodoptera exigua, Spodoptera exempta and Spodoptera litura.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

143.6 °F - closed cup

Flash Point(C)

62 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service