Skip to Content
Merck
All Photos(1)

Key Documents

SRP8044

Sigma-Aldrich

CD40L, Soluble human

recombinant, expressed in CHO cells, FLAG® tagged, >95% (SDS-PAGE)

Synonym(s):

ACRP30headless:CD154, ACRP30headless:CD40L, ACRP30headless:TNFSF5, ADIPOQ-CD40L

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
R$2,675.00

R$2,675.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
R$2,675.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.77

R$2,675.00


Check Cart for Availability

Request a Bulk Order

biological source

human

recombinant

expressed in CHO cells

tag

FLAG® tagged

Assay

>95% (SDS-PAGE)

form

lyophilized

mol wt

35-40 kDa by SDS-PAGE

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

<0.01 EU/μg endotoxin, tested

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CD40LG(959)

General description

CD40L (cluster of differentiation 40 ligand) belongs to the tumor necrosis factor (TNF) family, and is expressed by basophils, B cells, monocytic cells, natural killer cells, platelets, mast cells and T cells. It is predominantly expressed on pro-inflammatory T cells. It also exists in a soluble form.[1]

Biochem/physiol Actions

CD40L (cluster of differentiation 40 ligand) is active in its membrane-bound form.[1] It interacts with CD40 and functions as a proinflammatory and procoagulant agent. The serum levels of soluble CD40L is elevated in malignant middle cerebral artery infarction (MMCAI) patients, especially in non-surviving patients.[2]

Physical form

Lyophilized from PBS

Reconstitution

Reconstitute with 100 μL deionized water.

Other Notes

Human CD40L (aa 116-261) is fused at the N-terminus to mouse ACRP30headless (aa 18-111) and a FLAG®-tag.

Legal Information

FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Association between Serum Soluble CD154 Levels and Mortality in Patients with Malignant Middle Cerebral Artery Infarction.
Lorente L, et al.
International Journal of Molecular Sciences, 16(6), 12147-12158 (2015)
Two adjacent trimeric Fas ligands are required for Fas signaling and formation of a death-inducing signaling complex.
Holler N, et al.
Molecular and Cellular Biology, 23(4), 1428-1440 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service