Skip to Content
Merck
All Photos(1)

Key Documents

SRP8028

Sigma-Aldrich

DR6 (human): FC (human)

recombinant, expressed in HEK 293 cells, >95% (SDS-PAGE)

Synonym(s):

CD358, TNFRSF21, Tumor necrosis factor receptor superfamily member 21

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
R$1,441.00

R$1,441.00


Estimated to ship onMay 28, 2025



Select a Size

Change View
50 μG
R$1,441.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

R$1,441.00


Estimated to ship onMay 28, 2025


biological source

human

recombinant

expressed in HEK 293 cells

Assay

>95% (SDS-PAGE)

form

lyophilized

mol wt

~80 kDa by SDS-PAGE

packaging

pkg of 50 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

<0.1 EU/μg endotoxin, tested

color

white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

General description

DR6 is an orphan TNF receptor superfamily member belonging to a subgroup of receptors called death receptors. Expressed ubiquitously with high expression in lymphoid organs, heart, brain, and pancreas. Broadly expressed by developing neurons where it functions as pro-apoptotic factor. Recently, it has been reported that interaction with the beta-amyloid precursor protein (APP) activates a widespread caspase-dependent self-destruction program dependent on caspase-6.

Physical form

Lyophilized from PBS.

Reconstitution

Reconstitute with 50 μL deionized water. Further dilutions should be made with medium containing 5% fetal calf serum or a carrier protein.

Other Notes

The Fc portion of human IgG1 is fused to the C-terminus of human DR6 (aa 42-351).

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service