Skip to Content
Merck
All Photos(1)

Key Documents

SRP8011

Sigma-Aldrich

Granulin C human

recombinant, expressed in E. coli, His tagged, >90% (SDS-PAGE)

Synonym(s):

GRN C, Granulin-5

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
R$1,361.00

R$1,361.00


Check Cart for Availability


Select a Size

Change View
10 μG
R$1,361.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

R$1,361.00


Check Cart for Availability

biological source

human

recombinant

expressed in E. coli

tag

His tagged

Assay

>90% (SDS-PAGE)

form

liquid

mol wt

~10 kDa by SDS-PAGE

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

≥0.2 mg/mL

impurities

<1 EU/μg endotoxin, tested

color

clear

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... GRN(2896)

General description

Granulins have possible cytokine-like activity. They are important in normal development, inflammation, wound repair, and tumorigenesis.

Physical form

Solution in 55 mM TRIS-HCl, pH 8.2, containing 150 mM NaCl.

Other Notes

Human granulin C (aa 364-430) is fused at the N-terminus to a His-tag.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service