Skip to Content
Merck
All Photos(1)

Documents

SRP6354

Sigma-Aldrich

CD3E human

recombinant, expressed in E. coli, ≥90% (SDS-PAGE)

Synonym(s):

T-cell surface glycoprotein CD3 epsilon chain, T3E, TCRE

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥90% (SDS-PAGE)

form

liquid

mol wt

14.1 kDa (127 aa, 23-126 aa + His Tag)

packaging

pkg of 50 μg

concentration

1 mg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CD3E(916)

General description

CD3E is the CD3-epsilonpolypeptide, which together with CD3-gamma, -delta and -zeta, and the T-cell receptor α/β and gamma/delta heterodimers, forms the T-cell receptor-CD3 complex. This complex plays an important role in coupling antigen recognition to several intracellular signal-transduction pathways. The genes encoding the epsilon, gamma delta polypeptides are located in the same cluster on chromosome 11. The epsilon polypeptide plays an essential role in T-cell development. Defects in this gene cause immunodeficiency. This gene has also been linked to a susceptibility to type I diabetes in women. Recombinant human CD3E protein, fused to His-tag at N-terminus, was expressed in E.coli and purified by using conventional chromatography techniques.

Physical form

1 mg/mL in 20 mM Tris-HCl buffer (pH 8.0) containing 0.2 M NaCl, and 10% glycerol.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service