Skip to Content
Merck
All Photos(1)

Key Documents

SRP6330

Sigma-Aldrich

CD46 human

recombinant, expressed in E. coli, ≥85% (SDS-PAGE)

Synonym(s):

AHUS2, MCP, MIC10, Membrane cofactor protein isoform 14, TLX, TRA2.10

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥85% (SDS-PAGE)

form

liquid

mol wt

33.8 kDa (302 aa, 35-313 aa + His tag)

packaging

pkg of 100 μg

concentration

1 mg/mL

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CD46(4179)

General description

Membrane cofactor protein isoform 14, also known as CD46, is a type I membrane protein that has cofactor activity for inactivation of complement components C3b and C4b by serum factor I. This function resides in the complement control protein repeats (CCPs), with CCP1-4 essential for regulation. CD46 is widely distributed on thymocytes, T cells, B cells, monocytes, granulocytes, NK cells, platelets, endothelial cells, epithelial cells, fibroblasts, placenta and sperm, but not on erythrocyte. Recombinant human CD46 protein, fused to His-tag at N-terminus, was expressed in E.coli and purified by using conventional chromatography techniques.

Physical form

1 mg/mL in 20 mM Tris-HCl buffer (pH 8.0) containing 0.1 M NaCl and 20% glycerol.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi-Jun Jia et al.
Fish & shellfish immunology, 100, 317-323 (2020-03-17)
Largemouth bass ulcerative syndrome virus (LBUSV) is an important virus induce the mortality of largemouth bass (Micropterus Salmoides). In this study, we reported a single-walled carbon nanotubes (SWCNTs) containing LBUSV major capsid protein (MCP) subunit vaccine (SWCNTs-MCP) which was evaluated
Shao-Yeh Lu et al.
Applied and environmental microbiology, 85(11) (2019-03-25)
Microcin PDI (MccPDI), a class IIa microcin that is produced by Escherichia coli strains 25 and 284, is known to inhibit foodborne pathogenic enterohemorrhagic E. coli serotypes O157:H7 and O26. Here we demonstrate that MccPDI can inhibit Shigella strains and
Sinan Wang et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 27(5), 1305-1315 (2020-12-10)
We recently identified CD46 as a novel therapeutic target in prostate cancer. In this study, we developed a CD46-targeted PET radiopharmaceutical, [89Zr]DFO-YS5, and evaluated its performance for immunoPET imaging in murine prostate cancer models. [89Zr]DFO-YS5 was prepared and its in
Ying Yan et al.
Neuroscience letters, 701, 100-105 (2019-02-19)
P2X4 receptor (P2X4R), a subtype of P2 purinergic receptors, is an ATP-gated receptor through which activity of spinal microglia instigates pain hypersensitivity in various pain conditions. Accumulating evidence indicates that monocyte chemoattractant protein-1 (MCP-1) plays an important role in chronic
Jian Du et al.
Veterinary microbiology, 195, 87-95 (2016-10-25)
The outbreak of Chinese Giant Salamander (Andrias davidianus, CGS) Iridovirus (CGSIV) caused massive death of CGSs. However, some CGSs with low level of CGSIV usually could survive. In our study, major capsid protein (MCP) DNA replicates of CGSIV in shedding

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service