Skip to Content
Merck
All Photos(1)

Key Documents

SRP6169

Sigma-Aldrich

IL-1β human

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)

Synonym(s):

Catabolin, Endogenous pyrogen (EP), Leukocyte endogenous mediator (LEM), Lymphocyte-activating factor (LAF), Mononuclear cell factor (MCF)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

Assay

≥95% (SDS-PAGE)

form

lyophilized

mol wt

(18 and 25 kDa, monomer, glycosylated)

packaging

pkg of 10 μg

technique(s)

cell culture | mammalian: suitable

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... IL1B(3553)

General description

Interleukin-1β (IL1B) is produced by activated macrophages. It is synthesized as pro-IL-1β which is changed to active form IL-1β with the help of pro-inflammatory protease caspase-1. IL-1β belongs to the IL-1 gene family. It is a proinflammatory cytokine. The gene is mapped to human chromosome 2q14. In comparison with the Escherichia coli expressed protein, IL-1β is 50% more potent in promoting human CD4+ T cell differentiation into Th17 (T helper) cells. This cytokine is produced in a serum free, chemically defined media.

Application

  • IL-1β (IL1B) has been used to check the expression of NGAL (neutrophil gelatinase-associated lipocalin) in epithelial cells after stimulation with proinflammatory cytokines.
  • It has been used to check anti-inflammatory activities of N-Acetylglucosamine in human chondrocytes.
  • It has been used to create inflammatory endothelium in human coronary artery endothelial cells.

Biochem/physiol Actions

IL-1β (IL1B) stimulates thymocyte proliferation by inducing IL-2 release, B-cell maturation and proliferation and fibroblast growth factor activity. IL-1β proteins are involved in the inflammatory response, being identified as endogenous pyrogens, and are reported to stimulate the release of prostaglandin and collagenase from synovial cells. IL-1β is present in chronic inflamed tissues and also in tissues going for fibrogenesis. It brings neutrophil and macrophage to the site of infection. Mutations in IL-1β gene are linked with renal manifestations and renal sequelae in Henoch-Schönlein purpura. It is also associated with destruction of β-cell in type 1 diabetes.

Physical form

Lyophilized from a PBS solution.

Preparation Note

Centrifuge the vial prior to opening.

Reconstitution

Reconstitute in sterile PBS containing 0.1% endotoxin-free recombinant human serum albumin.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Proinflammatory cytokines detectable in synovial fluids from patients with temporomandibular disorders.
Takahashi T
Oral Surgery, Oral Medicine, Oral Pathology, Oral Radiology, and Endodontics, 85, 135-141 (1998)
Microbubbles targeted to intercellular adhesion molecule-1 bind to activated coronary artery endothelial cells.
Villanueva FS
Circulation, 98, 1-5 (1998)
Neutrophil gelatinase-associated lipocalin is up-regulated in human epithelial cells by IL-1 beta, but not by TNF-alpha.
Cowland JB
Journal of Immunology, 171, 6630-6639 (2003)
Autophagy controls IL-1beta secretion by targeting pro-IL-1beta for degradation.
Harris J
The Journal of Biological Chemistry, 286, 9587-9597 (2011)
N-acetylglucosamine prevents IL-1 beta-mediated activation of human chondrocytes.
Shikhman AR
Journal of Immunology, 166, 5155-5160 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service