Skip to Content
Merck
All Photos(1)

Key Documents

SRP6140

Sigma-Aldrich

hSUMO1-AMC human

≥95% (SDS-PAGE)

Synonym(s):

Small ubiquitin-related modifier 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

Assay

≥95% (SDS-PAGE)

form

lyophilized powder

mol wt

11.142

packaging

pkg of 50 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... SUMO1(7341)

General description

hSUMO1-AMC is prepared via the conjugation of 7-amino-4-methylcoumarin (AMC) to the C-terminal Gly of hSUMO1. This conjugation quenches the intrinsic fluorescence of AMC. Upon incubation with a protease recognizing hSUMOs, such as Senp1 or Senp2, AMC is released and the increase in fluorescence at 460 nm (Exc. 380 nm) can be measured. The enhanced purity of hSUMO1-AMC compared to that of other commercial vendors leads to a greater signal to noise ratio. This protein contains no extraneous tags.

Physical form

Lyophilized powder

Preparation Note

Centrifuge the vial prior to opening.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service