Skip to Content
Merck
All Photos(1)

Key Documents

SRP5349

Sigma-Aldrich

IKK-γ, GST tagged human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

AMCBX1, FIP-3, FIP3, Fip3p, IKBKG, IKK-gamma, IP, IP1, IP2, IPD2, NEMO

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
R$1,319.00

R$1,319.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Select a Size

Change View
20 μG
R$1,319.00

About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.32

R$1,319.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~73 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... IKBKG(8517)

General description

IKKgamma is a serine/threonine protein kinase that phosphorylates the I-kappa-B protein which is an inhibitor of the transcription factor NF-kappa-B complex. Phosphorylation of I-kappa-B protein triggers the degradation of the inhibitor via the ubiquitination pathway, thereby activating NF-kappa-B complex. IKKgamma forms dimers and trimers and interacts preferentially with IKKbeta but not IKKalpha. IKKgamma associates with activated ATM after the induction of DNA double-strand breaks. ATM phosphorylates serine-85 of IKKgamma to promote its ubiquitin-dependent nuclear export.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.

Preparation Note

After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service