Skip to Content
Merck
All Photos(2)

Key Documents

SRP5264

Sigma-Aldrich

KAT2A (GCN5), active, GST tagged human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

GCN5, GCN5L2, HsGCN5, MGC102791, PCAF-b, hGCN5

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
R$2,198.00

R$2,198.00


Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
10 μG
R$2,198.00

About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.32

R$2,198.00


Estimated to ship onJune 02, 2025


Request a Bulk Order

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

17.6-26.4 nmol/min·mg

mol wt

~82 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... KAT2A(2648)

General description

KAT2A (K (lysine) acetyltransferase 2A) is a histone acetyltransferase (HAT) that functions primarily as a transcriptional activator which functions as a repressor of NF-kappa-B by promoting ubiquitination of the NF-kappa-B subunit RELA in a HAT-independent manner. KAT2A control chromosome stability by coordinating the ATR checkpoint and double-strand break processing with autophagy. KAT2A acetyltransferases have homologous sequences and enzymatic activities which are important for recognition of nucleosomal substrates.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.

Preparation Note

After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service