Skip to Content
Merck
All Photos(1)

Key Documents

SRP5196

Sigma-Aldrich

I?Bβ , GST tagged human

recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

NFKBIB, TRIP9

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
R$2,849.00

R$2,849.00


Check Cart for Availability


Select a Size

Change View
50 μG
R$2,849.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

R$2,849.00


Check Cart for Availability

biological source

human

recombinant

expressed in E. coli

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~56 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... NFKBIB(4793)

General description

IκBβ is part of the NFκB complex and it inactivates NFκB complex by binding to it and trapping it in the cytoplasm. Phosphorylation of serine residues on IκBβ mediated by IκB kinases leads to its destruction via the ubiquitination pathway thereby allowing activation of the NFκB complex which translocates into the nucleus and binds DNA at kappa-B-binding motifs. The PEST domain of IkBβ can interact with two proteins KBRAS1 and KBRAS2 and this interaction can decrease the rate of degradation IκBβ. IκBβ has been shown to participate in multiple signaling pathways including adipocytokine signaling pathway, B-cell receptor signaling pathway, T-cell receptor signaling pathway and Hypoxia.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexander Hoffmann et al.
Science (New York, N.Y.), 298(5596), 1241-1245 (2002-11-09)
Nuclear localization of the transcriptional activator NF-kappaB (nuclear factor kappaB) is controlled in mammalian cells by three isoforms of NF-kappaB inhibitor protein: IkappaBalpha, -beta, and - epsilon. Based on simplifying reductions of the IkappaB-NF-kappaB signaling module in knockout cell lines
C Fenwick et al.
Science (New York, N.Y.), 287(5454), 869-873 (2000-02-05)
Small guanosine triphosphatases, typified by the mammalian Ras proteins, play major roles in the regulation of numerous cellular pathways. A subclass of evolutionarily conserved Ras-like proteins was identified, members of which differ from other Ras proteins in containing amino acids

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service