Skip to Content
Merck
All Photos(1)

Key Documents

SRP5147

Sigma-Aldrich

Troponin1, His tagged human

recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

DKFZp451O223, SSTNI, TNN1, TNNI1

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
R$2,309.00

R$2,309.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 μG
R$2,309.00

About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.32

R$2,309.00


Check Cart for Availability

Request a Bulk Order

biological source

human

recombinant

expressed in E. coli

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~24 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... TNNI1(7135)

General description

Troponin1 is a member of the tropomyosin family and regulates the calcium sensitivity of the myofibril contractile apparatus of striated muscles. Troponin1 is part of a complex with troponin T and troponin C in which Troponin1 acts as an inhibitory subunit that blocks actin-myosin interactions and thereby mediates striated muscle relaxation. Troponin1 is expressed in cardiac and skeletal muscle during early development but is restricted to slow-twitch skeletal muscle fibers in adults. The Troponin1 protein prevents muscle contraction by inhibiting calcium-mediated conformational changes in actin-myosin complexes.

Physical form

Supplied in 50mM sodium phosphate, pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.25mM DTT, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molecular basis of cardiac performance. Plasticity of the myocardium generated through protein isoform switches.
B Nadal-Ginard et al.
The Journal of clinical investigation, 84(6), 1693-1700 (1989-12-01)
N Tiso et al.
Biochemical and biophysical research communications, 230(2), 347-350 (1997-01-13)
In this paper the chromosomal localization of the human skeletal muscle genes Troponin-I slow-twitch (TNNI1), Troponin-I fast-twitch (TNNI2), and Troponin-C fast (TNNC2) and the refinement of the position for alpha-Tropomyosin (TPM1) and beta-Tropomyosin (TPM2) are reported. By radiation hybrid mapping

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service