Skip to Content
Merck
All Photos(1)

Key Documents

SRP5145

Sigma-Aldrich

TGFβR1 (80-end), GST tagged human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

AAT5, ACVRLK4, ALK-5, ALK5, SKR4, TGFR-1

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
R$2,038.00

R$2,038.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
20 μG
R$2,038.00

About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.32

R$2,038.00


Check Cart for Availability

Request a Bulk Order

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~65 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... TGFBR1(7046)

General description

TGFβR1 is a transmembrane serine/threonine protein kinase and a member of the TGFβ receptor subfamily. TGFβ regulates cell cycle progression by a unique signaling mechanism that involves its binding to TGFβR2 and activation of TGFβR1. TGFβR1 may be inactivated in many of the cases of human tumor cells refractory to TGFβ-mediated cell cycle arrest. Heterozygous mutations in TGFβR1 and TGFβR2 have been reported in Loeys-Dietz aortic aneurysm syndrome (LDS) and also dominant TGFβR2 mutations have been identified in Marfan syndrome type 2 (MFS2) and familial thoracic aortic aneurysms and dissections (TAAD). Mutations of TGFβR1 and TGFβR2 are associated with atherosclerosis and several human cancers.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gábor Mátyás et al.
Human mutation, 27(8), 760-769 (2006-06-23)
Very recently, heterozygous mutations in the genes encoding transforming growth factor beta receptors I (TGFBR1) and II (TGFBR2) have been reported in Loeys-Dietz aortic aneurysm syndrome (LDS). In addition, dominant TGFBR2 mutations have been identified in Marfan syndrome type 2
B K Suarez et al.
Prostate cancer and prostatic diseases, 8(1), 50-53 (2004-10-27)
The TGFBR1*6A (*6A) variant in exon 1 of the TGFBR1 gene has been postulated as a putative tumor susceptibility allele in several studies. We have performed a case-control study in 537 men with histologically verified prostate cancer and in 488

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service