Skip to Content
Merck
All Photos(1)

Key Documents

SRP5130

Sigma-Aldrich

SMAD1, GST tagged human

recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

BSP1, JV4-1, JV41, MADH1, MADR1

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
R$2,309.00

R$2,309.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 μG
R$2,309.00

About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.32

R$2,309.00


Check Cart for Availability

Request a Bulk Order

biological source

human

recombinant

expressed in E. coli

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~83 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... SMAD1(4086)

General description

SMAD1 is a member of the SMAD family which are signal transducers and transcriptional modulators that mediate multiple signaling pathways. The actions of bone morphogenetic proteins (BMPs) are mediated by SMAD1 and SMAD1 can be phosphorylated and activated by the BMP receptor kinase. Phosphorylated SMAD1 forms a complex with SMAD4 that is important for its function in the transcription regulation. The SMAD1-SMAD4 complex is a target for SMAD-specific E3 ubiquitin ligases, such as SMURF1 and SMURF2, and undergoes ubiquitination and proteasome-mediated degradation. The formation of a complex between STAT3 and SMAD1, bridged by p300, is involved in the cooperative signaling of LIF and BMP2 and the subsequent induction of astrocytes from neuronal progenitors.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P A Hoodless et al.
Cell, 85(4), 489-500 (1996-05-17)
Components of the signaling pathways that lie downstream of Ser/Thr kinase receptors and are required for signaling by the TGF beta superfamily have been poorly defined. The Drosophila gene Mothers against dpp (MAD) and the C. elegans sma genes are
K Nakashima et al.
Science (New York, N.Y.), 284(5413), 479-482 (1999-04-16)
The cytokines LIF (leukemia inhibitory factor) and BMP2 (bone morphogenetic protein-2) signal through different receptors and transcription factors, namely STATs (signal transducers and activators of transcription) and Smads. LIF and BMP2 were found to act in synergy on primary fetal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service