Skip to Content
Merck
All Photos(1)

Key Documents

SRP5126

Sigma-Aldrich

RGS1, His tagged human

recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

1R20, BL34, IER1, IR20

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

recombinant

expressed in E. coli

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~26 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... RGS1(5996)

General description

RGS1 is a member of the regulator of G-protein signalling family and attenuates the signalling activity of G-proteins. RGS1 binds to activated GTP-bound G alpha subunit and acts as a GTPase activating protein (GAP) thereby increasing the rate of conversion of GTP to GDP and terminating the signal. RGS1 is extensively up-regulated in renal cell carcinoma (RCC) tissues and melanoma. In melanoma, RGS1 expression is significantly correlated with increased tumor thickness, mitotic rate and presence of vascular involvement. Furthermore, there is significant association between increasing RGS1 expression and reduced relapse-free survival as well as disease-specific survival (DSS) survival.

Physical form

Supplied in 50mM sodium phosphate, pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.25mM DTT, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David A Sierra et al.
Genomics, 79(2), 177-185 (2002-02-07)
The regulators of G-protein signaling (RGS) proteins are important regulatory and structural components of G-protein coupled receptor complexes. RGS proteins are GTPase activating proteins (GAPs) of Gi-and Gq-class Galpha proteins, and thereby accelerate signaling kinetics and termination. Here, we mapped
Javier Rangel et al.
The American journal of surgical pathology, 32(8), 1207-1212 (2008-06-27)
RGS1 (regulator of G protein signaling 1) encodes a member of the regulator of G protein family. Recently, RGS1 was found to be overexpressed in gene expression-profiling studies of melanoma. However, no analyses have been reported of its expression at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service