Skip to Content
Merck
All Photos(1)

Key Documents

SRP5115

Sigma-Aldrich

P73g, GST tagged human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

TAp73γ, TP73

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
R$2,097.00

R$2,097.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 μG
R$2,097.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

R$2,097.00


Check Cart for Availability

Request a Bulk Order

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~85 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... TP73(7161)

General description

p73γ is a member of the p53 family of transcription factors which are involved in cellular responses to stress and development. There are four isoforms of p73 (p73α, p73β, p73δ and p73γ) which differed in their abilities to form homodimeric and heterodimeric interactions with each other and with p53. The p73 isoforms also differed in their abilities to activate transcription of the p21(Waf1) promoter and to inhibit colony formation. The p73 protein is expressed at very low levels in normal tissues and is differentially expressed in a number of tumors. p73 is a stress-response gene that activates transcription of p53-responsive genes and inhibits cell growth in a p53-like manner by inducing apoptosis.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V De Laurenzi et al.
The Journal of experimental medicine, 188(9), 1763-1768 (1998-11-06)
p73 has been recently identified as a new structural and functional homologue of the transcription factor p53. It is expressed in either a full-length form, alpha, or a shorter beta mRNA variant, with exon 13 spliced out. Here we report
C A Jost et al.
Nature, 389(6647), 191-194 (1997-09-20)
The protein p53 is the most frequently mutated tumour suppressor to be identified so far in human cancers. The ability of p53 to inhibit cell growth is due, at least in part, to its ability to bind to specific DNA

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service