SRP5110
P300 (1283-1673), GST tagged human
recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Synonym(s):
EP300, KAT3B
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
human
recombinant
expressed in baculovirus infected Sf9 cells
Assay
≥70% (SDS-PAGE)
form
buffered aqueous glycerol solution
mol wt
~74 kDa
NCBI accession no.
application(s)
cell analysis
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... EP300(2033)
General description
P300 encodes the adenovirus E1A-associated cellular p300 transcriptional co-activator protein. P300 is related by sequence to CBP and like CBP can stimulate transcription through activation of CREB. The P300 activity is specifically inhibited by the adenovirus oncoprotein E1A. P300 has also been identified as a co-activator of HIF1A (hypoxia-inducible factor 1 alpha), and thus plays a role in the stimulation of hypoxia-induced genes such as VEGF. P300 is a components of DRAF1 (double-stranded RNA-activated factor-1), a positive regulator of interferon-stimulated gene transcription that functions as a direct response to viral infection.
Physical form
Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.
Preparation Note
after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Molecular and cellular biology, 18(3), 1359-1368 (1998-03-06)
Cells respond to viral infection or double-stranded RNA with the transcriptional induction of a subset of alpha/beta interferon-stimulated genes by a pathway distinct from the interferon signal pathway. The transcriptional induction is mediated through a DNA sequence containing the alpha/beta
Genes & development, 8(8), 869-884 (1994-04-15)
The growth-controlling functions of the adenovirus E1A oncoprotein depend on its ability ot interact with a set of cellular proteins. Among these are the retinoblastoma protein, p107, p130, and p300. We have isolated a cDNA encoding full-length human p300 and
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service