Skip to Content
Merck
All Photos(1)

Key Documents

SRP5107

Sigma-Aldrich

p19INK4D, GST tagged human

recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

CDKN2D, INK4D, p19, p19-INK4D

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
R$2,850.00

R$2,850.00


Estimated to ship onMay 28, 2025



Select a Size

Change View
50 μG
R$2,850.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

R$2,850.00


Estimated to ship onMay 28, 2025


recombinant

expressed in E. coli

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~44 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CDKN2D(1032)

General description

p19INK4D protein is associated with CDK6 in several hematopoietic cell lines. It has been demonstrated that p19INK4D expression enhances cell survival under genotoxic conditions. By using p19INK4D-overexpressing cells, it has been demonstrated that p19INK4D expression correlates with the cellular resistance to UV treatment with increased DNA repair activity against UV-induced lesions. p19INK4D regulates protein network that integrate DNA repair, apoptosis and checkpoint mechanisms in order to maintain the genomic integrity.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julieta M Ceruti et al.
Oncogene, 24(25), 4065-4080 (2005-03-08)
The genetic instability driving tumorigenesis is fueled by DNA damage and by errors made by the DNA replication. Upon DNA damage the cell organizes an integrated response not only by the classical DNA repair mechanisms but also involving mechanisms of
K L Guan et al.
Molecular biology of the cell, 7(1), 57-70 (1996-01-01)
Cyclin-dependent kinases 4 and 6 are complexed with many small cellular proteins in vivo. We have isolated cDNA sequences, INK4d, encoding a 19-kDa protein that is associated with CDK6 in several hematopoietic cell lines. p19 shares equal similarity and a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service