Skip to Content
Merck
All Photos(2)

Key Documents

SRP5100

Sigma-Aldrich

YES1, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

HsT441, P61-YES, Yes, c-yes

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
R$3,062.00

R$3,062.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
R$3,062.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

R$3,062.00


Check Cart for Availability

Request a Bulk Order

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

157-213 nmol/min·mg

mol wt

~91 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... YES1(7525)

General description

YES1 is the cellular homolog of the Yamaguchi sarcoma virus oncogene that has tyrosine kinase activity and belongs to the SRC family. YES1 lies in close proximity to thymidylate synthase gene on chromosome 18 and chromosome 22. The activation of YES1 may play a significant role in the malignant transformation of hepatocytes, and is important for maintaining embryonic stem cells in an undifferentiated state. YES1 is a useful marker to detect early-stage hepatocellular carcinoma, and it plays a key role in the tumorigenesis and metastasis of gastric cancer. YES1 induction results in increased cancer cell motility suggesting that YES1 may promote cancer spread and metastasis rather than tumor growth.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G A Silverman et al.
Genomics, 15(2), 442-445 (1993-02-01)
The YES1 proto-oncogene was mapped previously to human chromosome band 18q21.3 by using isotopic in situ hybridization. Using yeast artificial chromosomes (YACs) as probes and fluorescence in situ hybridization, a strong signal was detected in the region corresponding to 18p11.3.
Jane Barraclough et al.
Neoplasia (New York, N.Y.), 9(9), 745-754 (2007-09-28)
Increases in the levels and/or activity of nonreceptor tyrosine kinases c-Src and c-Yes are often associated with colorectal carcinogenesis. The physiological consequences of increased c-Yes activity during the early and late stages of tumorigenesis, in addition to the degree of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service