Skip to Content
Merck
All Photos(2)

Key Documents

SRP5083

Sigma-Aldrich

RIPK5, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

DSTYK, DustyPK, HDCMD38P, KIAA0472, RIP5

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
R$3,062.00

R$3,062.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
R$3,062.00

About This Item

UNSPSC Code:
51111800
NACRES:
NA.32

R$3,062.00


Check Cart for Availability

Request a Bulk Order

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

5-7 nmol/min·mg

mol wt

~140 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... DSTYK(25778)

General description

RIPK5 is a dual specificity serine/threonine and tyrosine protein kinase that is a member of the RIPK family. RIPK5 is widely expressed in vertebrates with broad distributed in the central nervous system, and deregulated in certain human cancers. RIPK5 has been shown to function as a regulator of cell death. Overexpression of RIPK5 leads to cell death as evidenced by DNA fragmentation. RIPK5 induces both caspase-dependent and caspase-independent cell death, and N- and C-terminal RIPK5 deletion mutants retained the ability to induce cell death.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jikun Zha et al.
Biochemical and biophysical research communications, 319(2), 298-303 (2004-06-05)
Members of the RIP serine/threonine kinase family are involved in activation of NF-kappaB, JNK, and p38, and induction of apoptosis. Here we report the identification of a novel RIP-homologous protein designated as RIP5. The C-terminus of RIP5 contains a kinase
Jianbin Peng et al.
Biochimica et biophysica acta, 1759(11-12), 562-572 (2006-11-25)
Ser/Thr- and Tyr-Protein kinases constitute a key switch underlying the dynamic nature and graded regulation of signal transduction and pathway activities in cellular organization. Here we describe the identification and characterization of Dusty, a single-copy gene that arose in metazoan

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service