Skip to Content
Merck
All Photos(2)

Key Documents

SRP5066

Sigma-Aldrich

PIM1, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

PIM

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
R$4,083.00

R$4,083.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

Select a Size

Change View
10 μG
R$4,083.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

R$4,083.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

269-363 nmol/min·mg

mol wt

~62 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PIM1(5292)

General description

PIM1 is a proto-oncogene that belongs to a family of serine/threonine protein kinases that are highly conserved through evolution in multicellular organisms. Originally identified from Moloney murine leukemia virus induced T-cell lymphomas in mice, PIM1 is involved in the control of cytokine-mediated cell proliferation, differentiation and survival of lymphoid and myeloid cells as well as others. Expression of PIM1 can be stimulated by a variety of growth factors and is regulated at four different levels: transcriptional, post-transcriptional, translational and post-translational. Expression of PIM1 is mediated through activation of the JAK/STAT pathway.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Friedmann et al.
Archives of biochemistry and biophysics, 298(2), 594-601 (1992-11-01)
The human pim-1 proto-oncogene was expressed in Escherichia coli as a glutathione-S-transferase (GST)-fusion protein and the enzymatic properties of its kinase activity were characterized. Likewise, a Pim-1 mutant lacking intrinsic kinase activity was constructed by site-directed mutagenesis (Lys67 to Met)
T C Meeker et al.
Journal of cellular biochemistry, 35(2), 105-112 (1987-10-01)
The mouse PIM-1 gene has been implicated in the evolution of retrovirus-associated mouse lymphomas. We have initiated a study of the human PIM-1 gene because of its potential importance as a human oncogene. We have isolated genomic and cDNA clones

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service