Skip to Content
Merck
All Photos(2)

Key Documents

SRP5051

Sigma-Aldrich

NEK11, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

FLJ23495

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
R$3,152.00

R$3,152.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
R$3,152.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

R$3,152.00


Check Cart for Availability

Request a Bulk Order

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

21-29 nmol/min·mg

mol wt

~110 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... NEK11(79858)

General description

NEK11 is a member of the NEK family and the protein localizes to the nucleoli where it may function with NEK2A in the S-phase checkpoint. NEK2 phosphorylates NEK11 in its C-terminal autoinhibitory domain and elevates NEK11 kinase activity by dissociating the autoinhibitory domain from the N-terminal catalytic domain. NEK11 protein appears to play a role in DNA replication and response to genotoxic stress. NEK11 activity is activated in HeLa cells treated with various DNA-damaging agents and replication inhibitors. Activation of NEK11 by genotoxic agents is suppressed by inhibitor of ATR/ATM-mediated signalling indicating that NEK11 has a role in S-phase checkpoint downstream of ATR/ATM signalling.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kohji Noguchi et al.
The Journal of biological chemistry, 277(42), 39655-39665 (2002-08-03)
DNA replication and genotoxic stresses activate various checkpoint-associated protein kinases, and checkpoint dysfunction often leads to cell lethality. Here, we have identified new members of the mammalian NIMA family of kinases, termed Nek11L and Nek11S (NIMA-related kinase 11 Long and
Kohji Noguchi et al.
The Journal of biological chemistry, 279(31), 32716-32727 (2004-05-27)
We previously reported that Nek11, a member of the NIMA (never-in-mitosis A) family of kinases, is activated in G(1)/S-arrested cells. We provide herein several lines of evidence for a novel interaction between Nek11 and Nek2A. Both Nek11 and Nek2A, but

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service