Skip to Content
Merck
All Photos(1)

Documents

SRP4174

Sigma-Aldrich

IL-16 from mouse

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)

Synonym(s):

IL16, KIAA4048, LCF, Lymphocyte Chemoattractant Factor, mKIAA4048, prIL-16

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

mouse

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

mol wt

~13.2 kDa

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

endotoxin, tested

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Il16(16170)

General description

Interleukin 16 (IL-16), also known as lymphocyte chemoattractant factor (LCF), stimulates a migratory response in CD4+ T cells and CD4+ monocytes and eosinophils. In addition, human IL-16 induces expression of the IL-2 receptor by T cells. mouse IL-16 is a 13.2 kDa protein containing 127 amino acid residues.

Physical form

Lyophilized from 5 mM Sodium Phosphate, pH 7.6.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitute in water to a concentration of 0.1-1 mg/mL. The solution can then be diluted into other aqueous buffers.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service