Skip to Content
Merck
All Photos(1)

Key Documents

SRP4163

Sigma-Aldrich

IL-12 from rat

recombinant, expressed in baculovirus infected Sf9 cells, ≥95% (SDS-PAGE), ≥95% (HPLC)

Synonym(s):

CTL maturation factor (TCMF), Cytotoxic lymphocyte maturation factor (CLMF), Edodekin-α, IL-12, NKSF, TSF

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

rat

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥95% (HPLC)
≥95% (SDS-PAGE)

form

lyophilized

potency

0.01-0.05 ng/mL

mol wt

~70 kDa

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

endotoxin, tested

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

General description

The recombinant rat IL-12 produced from Sf9 insect cells is a glycosylated disulfide linked hetromeric polypeptide containing 503 amino acids and having a molecular weight of 70 kDa comprised of disulfide-bonded 35 kDa and 40 kDa subunits

Physical form

Lyophilized from 1 mg/ml solution with no additives.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitute in water to a concentration of 0.1-0.5 mg/mL (Note: Gently pipetting to dissolve). The solution can be diluted into other aqueous buffers.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service