Skip to Content
Merck
All Photos(1)

Key Documents

SRP3301

Sigma-Aldrich

MIDKINE mouse

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)

Synonym(s):

MK, NEGF-2

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
R$1,758.00

R$1,758.00


Estimated to ship onMay 25, 2025



Select a Size

Change View
20 μG
R$1,758.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

R$1,758.00


Estimated to ship onMay 25, 2025


biological source

mouse

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

mol wt

13.4 kDa

packaging

pkg of 20 μg

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... MDK(17242)

General description

MK plays an important regulatory role in epithelial-mesenchymal interactions during fetal development and in postnatal lung development. MK chemoattracts embryonic neurons, neutrophils and macrophages, and by signaling through the ALK receptor it exerts angiogenic, growth and survival activities during tumorgenesis.

Biochem/physiol Actions

Midkine and its functionally-related protein Pleiotrophin are heparin-binding neurotrophic factors that signal through the same receptor, known as anaplastic lymphoma kinase (ALK). Recombinant murine Midkine is a 13.4 kDa protein containing 123 amino acid residues including five intra-molecular disulfide bonds.

Physical form

Lyophilized from 0.4x PBS, pH 7.4.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W Iwasaki et al.
The EMBO journal, 16(23), 6936-6946 (1998-01-31)
Midkine (MK) is a 13 kDa heparin-binding polypeptide which enhances neurite outgrowth, neuronal cell survival and plasminogen activator activity. MK is structurally divided into two domains, and most of the biological activities are located on the C-terminal domain. The solution
Neibla Priego et al.
Nature medicine, 24(7), 1024-1035 (2018-06-13)
The brain microenvironment imposes a particularly intense selective pressure on metastasis-initiating cells, but successful metastases bypass this control through mechanisms that are poorly understood. Reactive astrocytes are key components of this microenvironment that confine brain metastasis without infiltrating the lesion.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service