Skip to Content
Merck
All Photos(1)

Key Documents

SRP3162

Sigma-Aldrich

sTNF-receptor Type I human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

Soluble Tumor Necrosis Factor type I, TNF-R55, TNFAR, TNFRSF1A

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
R$2,796.00

R$2,796.00


Estimated to ship onApril 07, 2025


Request a Bulk Order

Select a Size

Change View
20 μG
R$2,796.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

R$2,796.00


Estimated to ship onApril 07, 2025


Request a Bulk Order

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

0.25 ng/mL ED50

mol wt

18.3 kDa

packaging

pkg of 20 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... TNFRSF1A(7132)

General description

TNFR1 (tumor necrosis factor receptor 1) gene is localized to human chromosome 12p13, and encodes a 55kDa receptor showing a wide range of tissue expression. This receptor belongs to the family of receptors which contain repeating cysteine-rich extracellular motifs. TNFR1 contains a protein-protein interacting domain called death domain containing ∼70 amino acids. It also contains a silencer of death domains (SODD), a 60kDa domain, which inhibits ligand-independent self-association of TNFR1.[1] TNFR1 is a member of the TNFR family, which are type I transmembrane proteins.[2]
Recombinant human sTNFR1 is an 18.3 kDa protein (162 amino acid residues) comprising the cysteine rich ligand binding portion of the extracellular domain of the TNFR1 protein.

Biochem/physiol Actions

TNFR1 (tumor necrosis factor receptor 1) is involved in inflammatory responses, and the cleavage of the membrane form leads to production of soluble TNFR1. The soluble form competes with the membrane-bound form, and this leads to a reduction in the severity of the inflammatory responses. Mutations in this gene resulting in the disruption of conserved extracellular disulfide bonds, leads to lower levels of soluble TNFR1 forms, which is linked with autosomal dominant periodic fever syndromes, characterized by unexplained recurrent fevers and severe localized inflammation.[1] Studies in two independent community-based elderly cohorts show that sTNFR1 is linked with increased mortality in cardiovascular and cancer cases.[3]

Sequence

MDSVCPQGKY IHPQNNSICC TKCHKGTYLY NDCPGPGQDT DCRECESGSF TASENHLRHC LSCSKCRKEM GQVEISSCTV DRDTVCGCRK NQYRHYWSEN LFQCFNCSLC LNGTVHLSCQ EKQNTVCTCH AGFFLRENEC VSCSNCKKSL ECTKLCLPQI EN

Physical form

Lyophilized from 10 mM Tris, pH 8.0 + 20 mM NaCl.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Soluble tumor necrosis factor receptor 1 (sTNFR1) is associated with increased total mortality due to cancer and cardiovascular causes - findings from two community based cohorts of elderly.
Carlsson AC et al
Atherosclerosis, 237(1), 236-242 (2014)
Signalling pathways of the TNF superfamily: a double-edged sword.
Aggarwal BB
Nature Reviews: Immunology, 3(9), 745-756 (2003)
Germline mutations in the extracellular domains of the 55 kDa TNF receptor, TNFR1, define a family of dominantly inherited autoinflammatory syndromes.
McDermott MF et al
Cell, 97(1), 133-144 (1999)
Aggarwal, B.B., et al.
Cytokine Reference, 1619-1632 (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service