Skip to Content
Merck
All Photos(1)

Documents

SRP3106

Sigma-Aldrich

LIGHT human

recombinant, expressed in Hi-5 Insect cells, ≥96% (SDS-PAGE), ≥96% (HPLC), suitable for cell culture

Synonym(s):

HVEM-L, TNFSF14

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in Hi-5 Insect cells

Assay

≥96% (HPLC)
≥96% (SDS-PAGE)

form

lyophilized

mol wt

20.0-22.5 kDa

packaging

pkg of 15 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... TNFSF14(8740)

General description

LIGHT belongs to the TNF family of ligands, and can signal through the herpes virus entry mediator type A receptor (HVEM, TNFRSF14), LTbR, or bind to a decoy receptor, DcR3. It is expressed in splenocytes, activated PBL, CD+8 tumor infiltrating lymphocytes, granulocytes, and monocytes. LIGHT has the ability to active NFkB, to co-stimulate the activation of lymphocytes and to induce apoptosis in certain human tumor cells. Recombinant human LIGHT has a calculated mass of 19.3 kDa containing 177 amino acid residues. Due to glycosylation LIGHT migrates between 20.0-22.5 kDa by SDS-PAGE under non-reducing conditions.

Biochem/physiol Actions

LIGHT belongs to the TNF family of ligands, and can signal through the herpes virus entry mediator type A receptor (HVEM, TNFRSF14), LTbR, or bind to a decoy receptor, DcR3. Recombinant human LIGHT has a calculated mass of 19.3 kDa containing 177 amino acid residues. Due to glycosylation LIGHT migrates between 20.0-22.5 kDa by SDS-PAGE under non-reducing conditions.

Sequence

RLGEMVTRLP DGPAGSWEQL IQERRSHEVN PAAHLTGANS SLTGSGGPLL WETQLGLAFL RGLSYHDGAL VVTKAGYYYI YSKVQLGGVG CPLGLASTIT HGLYKRTPRY PEELELLVSQ QSPCGRATSS SRVWWDSSFL GGVVHLEAGE KVVVRVLDER LVRLRDGTRS YFGAFMV

Physical form

Lyophilized from 20 mM Sodium Phosphate, pH 7.0.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huai Na Qiu et al.
Mediators of inflammation, 2014, 136463-136463 (2014-05-02)
Allergic asthma can cause airway structural remodeling, involving the accumulation of extracellular matrix and thickening of smooth muscle. Tumor necrosis factor (TNF) family ligand LIGHT (TNFSF14) is a cytokine that binds herpesvirus entry mediator (HVEM)/TNFRSF14 and lymphotoxin β receptor (LTβR).
Rana Herro et al.
The Journal of investigative dermatology, 135(8), 2109-2118 (2015-03-20)
Several inflammatory diseases including scleroderma and atopic dermatitis display dermal thickening, epidermal hypertrophy, or excessive accumulation of collagen. Factors that might promote these features are of interest for clinical therapy. We previously reported that LIGHT, a TNF superfamily molecule, mediated
Yu Mikami et al.
PloS one, 9(12), e114791-e114791 (2014-12-17)
Asthma-related mortality has been decreasing due to inhaled corticosteroid use, but severe asthma remains a major clinical problem. One characteristic of severe asthma is resistance to steroid therapy, which is related to neutrophilic inflammation. Recently, the tumor necrosis factor superfamily
Bianca Crifo et al.
Journal of immunology (Baltimore, Md. : 1950), 202(5), 1521-1530 (2019-02-01)
Hypoxia is a common and prominent feature of the microenvironment at sites of bacteria-associated inflammation in inflammatory bowel disease. The prolyl-hydroxylases (PHD1/2/3) and the asparaginyl-hydroxylase factor-inhibiting HIF are oxygen-sensing enzymes that regulate adaptive responses to hypoxia through controlling the activity
Sook-Kyoung Heo et al.
Experimental & molecular medicine, 44(2), 149-158 (2011-11-25)
The development of gastric cancer (GC) is closely related to chronic inflammation caused by Helicobacter pylori infection, and herpes virus entry mediator (HVEM) is a receptor expressed on the surface of leukocytes that mediates potent inflammatory responses in animal models.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service