Skip to Content
Merck
All Photos(1)

Key Documents

SRP3054

Sigma-Aldrich

HCC-1 Peptide

≥98% (HPLC), human recombinant, expressed in E. coli, suitable for cell culture, 72 aa

Synonym(s):

CCL14, HCC-1/HCC-3, Hemofiltrate CC Chemokine-1, NCC-2

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
R$2,850.00

R$2,850.00


Check Cart for Availability


Select a Size

Change View
10 μG
R$2,850.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

R$2,850.00


Check Cart for Availability

Product Name

HCC-1 (CCL14) (72 aa) human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

5.0-20.0 ng/mL

mol wt

7.9 kDa

packaging

pkg of 10 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CCL14(6358)

General description

HCC-1 is a CC chemokine that signals through the CCR1 receptor and chemoattracts blood monocytes. It is secreted by various tissues including skeletal muscle, heart, spleen, liver and bone marrow. Recombinant human HCC-1 is an 8.4 kDa protein consisting of 72 amino acids including the four highly conserved residues present in CC chemokines.

Biochem/physiol Actions

HCC-1 is a CC chemokine that signals through the CCR1 receptor and chemoattracts blood monocytes. Recombinant human HCC-1 is an 8.4 kDa protein consisting of 72 amino acids including the four highly conserved residues present in CC chemokines.

Sequence

TESSSRGPYH PSECCFTYTT YKIPRQRIMD YYETNSQCSK PGIVFITKRG HSVCTNPSDK WVQDYIKDMK EN

Physical form

Lyophilized with no additives.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C L Tsou et al.
The Journal of experimental medicine, 188(3), 603-608 (1998-08-04)
Hemofiltrate C-C chemokine (HCC)-1 is a recently cloned C-C chemokine that is structurally similar to macrophage inflammatory protein (MIP)-1alpha. Unlike most chemokines, it is constitutively secreted by tissues and is present at high concentrations in normal human plasma. Also atypical
P Schulz-Knappe et al.
The Journal of experimental medicine, 183(1), 295-299 (1996-01-01)
A novel CC chemokine, HCC-1, was isolated from the hemofiltrate of patients with chronic renal failure. HCC-1 has a relative molecular mass of 8,673 and consists of 74 amino acids including four cysteines linked to disulfide bonds. HCC-1 cDNA was

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service