Skip to Content
Merck
All Photos(1)

Key Documents

SRP2116

Sigma-Aldrich

RNA Polymerase II, p15.6 subunit, GST tagged human

recombinant, expressed in E. coli, ≥85% (SDS-PAGE)

Synonym(s):

HRBP14.4, POLRF, RPABC2, RPB14.4, RPB6

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
R$3,295.00

R$3,295.00


Estimated to ship onJune 02, 2025



Select a Size

Change View
10 μG
R$3,295.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.26

R$3,295.00


Estimated to ship onJune 02, 2025


biological source

human

recombinant

expressed in E. coli

Assay

≥85% (SDS-PAGE)

form

frozen liquid

mol wt

~40 kDa

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

200 μg/mL

color

clear colorless

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... POLR2F(5435)

Biochem/physiol Actions

hRPB6 is a highly conserved subunit shared by all three RNA polymerases, consisting of 142 amino acid residues. The gene for yeast RPB6 is essential for cell viability and homologues of this subunit exist in archaeal and some viral RNA polymerases. For example the bacterial w (the fifth subunit of the bacterial RNAP core enzyme) and eukaryotic RPB6 are structural homologs. RPB6 has been shown to promote pol II complex assembly, and/or increase its stability, through specific interactions with the largest subunit of RNA pol II. In addition, RPB6 was found to make contact with three small subunits, RPB5, RPB7, and RPB8 and to play a role in the interaction between RNA polymerase II and the transcription elongation factor TFIIS.

Physical form

Clear and colorless frozen liquid solution

Preparation Note

Use a manual defrost freezer and avoid repeated freeze-thaw cycles. While working, please keep sample on ice.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Ishiguro et al.
Molecular and cellular biology, 20(4), 1263-1270 (2000-01-29)
The Rpb6 subunit of RNA polymerase II is one of the five subunits common to three forms of eukaryotic RNA polymerase. Deletion and truncation analyses of the rpb6 gene in the fission yeast Schizosaccharomyces pombe indicated that Rpb6, consisting of
L Minakhin et al.
Proceedings of the National Academy of Sciences of the United States of America, 98(3), 892-897 (2001-02-07)
Bacterial DNA-dependent RNA polymerase (RNAP) has subunit composition beta'betaalpha(I)alpha(II)omega. The role of omega has been unclear. We show that omega is homologous in sequence and structure to RPB6, an essential subunit shared in eukaryotic RNAP I, II, and III. In
N A Woychik et al.
Genes & development, 4(3), 313-323 (1990-03-01)
RNA polymerases I, II, and III share three subunits that are immunologically and biochemically indistinguishable. The Saccharomyces cerevisiae genes that encode these subunits (RPB5, RPB6, and RPB8) were isolated and sequenced, and their transcriptional start sites were deduced. RPB5 encodes

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service